Learn More
Medchemexpress LLC Thrombin (MW 37kDa) | 9002-04-4 | 37000 (Average) | 2000 U
SDP
Catalog No. 5000435970
Encompass_Preferred
Thrombin (MW 37kDa) is a Na+-activated, allosteric serine protease that plays opposing functional roles in blood coagulation. It has a Thrombin recognition sequence and can be used to digest GST-tagged proteins.
  • Na+-activated, allosteric serine protease.
  • Plays opposing functional roles in blood coagulation.
  • Can be used to digest GST-tagged proteins.
Catalog No. 50-004-35970 Supplier Medchemexpress LLC Supplier No. HY1141642000U
Thrombin (MW 37kDa) is a Na+-activated, allosteric serine protease that plays opposing functional roles in blood coagulation. It has a Thrombin recognition sequence and can be used to digest GST-tagged proteins.
  • Na+-activated, allosteric serine protease.
  • Plays opposing functional roles in blood coagulation.
  • Can be used to digest GST-tagged proteins.