Learn More
Sigma Aldrich Fine Chemicals Biosciences Thrombin Protease Cytiva 2
Catalog No. 502901681
Encompass
Thrombin is a protease used to digest fusion proteins prepared from pGEX vectors recognition sequence for thrombin (pGEX-1lT pGEX-2T pGEX-2TK pGEX-4T-1 pGEX-4T-2 and pGEX-4T-3). Purified from bovine plasma.Thrombin is a protease used to digest fusion proteins prepared from pGEX vectors (see pGEX Vectors (GST Gene fusion System)) containing the recognition sequence for thrombin (pGEX-1lT pGEX-2T pGEX-2TK pGEX-4T-1 pGEX-4T-2 and pGEX-4T-3).
Catalog No. 50-290-1681 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE27084601
Thrombin is a protease used to digest fusion proteins prepared from pGEX vectors recognition sequence for thrombin (pGEX-1lT pGEX-2T pGEX-2TK pGEX-4T-1 pGEX-4T-2 and pGEX-4T-3). Purified from bovine plasma.Thrombin is a protease used to digest fusion proteins prepared from pGEX vectors (see pGEX Vectors (GST Gene fusion System)) containing the recognition sequence for thrombin (pGEX-1lT pGEX-2T pGEX-2TK pGEX-4T-1 pGEX-4T-2 and pGEX-4T-3).