Learn More
Medchemexpress LLC Thrombin Receptor/PA 50ug
SDP
Catalog No. 5000180729
Encompass_Preferred
The thrombin receptor/PAR1 protein is a high-affinity receptor for activated thrombin that is coupled to G proteins to stimulate phosphoinositide hydrolysis This signaling mechanism suggests its involvement in platelet activation which is critical for hemostasis and thrombosis Thrombin Receptor/PAR1 Protein Human (HEK293 His) is the recombinant human-derived Thrombin Receptor/PAR1 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80729 Supplier Medchemexpress LLC Supplier No. HYP7361250UG
The thrombin receptor/PAR1 protein is a high-affinity receptor for activated thrombin that is coupled to G proteins to stimulate phosphoinositide hydrolysis This signaling mechanism suggests its involvement in platelet activation which is critical for hemostasis and thrombosis Thrombin Receptor/PAR1 Protein Human (HEK293 His) is the recombinant human-derived Thrombin Receptor/PAR1 protein expressed by HEK293 with C-His labeled tag