Learn More
Cayman Chemical ThrombIn RECPTR PeptID LIg 1mg
SDP
Catalog No. 502934227
Encompass_Preferred
An antagonist of the thrombin receptor (EC50s = 16-33 µM); inhibits α-thrombin and TRAP-induced platelet aggregation in vitro at 32 µM; inhibits arterial thrombosis in a rabbit model of partial carotid artery occlusion at 100 µmol/kg bolus, i.v., plus 900 µmol/kg infusion)
Catalog No. 50-293-4227 Supplier Cayman Chemical Supplier No. 249951 MG
An antagonist of the thrombin receptor (EC50s = 16-33 µM); inhibits α-thrombin and TRAP-induced platelet aggregation in vitro at 32 µM; inhibits arterial thrombosis in a rabbit model of partial carotid artery occlusion at 100 µmol/kg bolus, i.v., plus 900 µmol/kg infusion)