Learn More
Haematologic Technologies Inc THROMBIN SUBSTRATE 1MG
Supplier:  Haematologic Technologies Inc SN20
Encompass_Preferred
Fluorogenic substrate, Contains the fluorescent reporter group 6-amino-1-naphthalene-sulfonamide (ANSN) for monitoring enzyme activity of the serine protease THROMBIN, Boc-L-FPR-ANSNH-C2H5, 1 milligram, RUO, Supplied in DMSO, store at -20 Celsius.
Catalog No. 50-883-412