Learn More
CPC Scientific Benzoyl-Phe-Val-Arg-AMC (trifluoroacetate salt) 25MG
SDP
Catalog No. 502109806
Encompass
ONE-LETTER SEQUENCE: Benzoyl-FVR-AMCMOLECULAR FORMULA: C37H43N7O6MOLECULAR WEIGHT: 681.79STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [88899-22-3]SYNONYMS: Bz-Phe-Val-Arg-AMCRESEARCH AREA: HematologyREFERENCES:L. Svendsen, et al., Folia Haematol, 98, 446 (1972)R. Lottenberg, et al., Meth. Enz., 80, 341 (1981)
Catalog No. 50-210-9806 Supplier CPC Scientific Supplier No. MMPS027C
ONE-LETTER SEQUENCE: Benzoyl-FVR-AMCMOLECULAR FORMULA: C37H43N7O6MOLECULAR WEIGHT: 681.79STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [88899-22-3]SYNONYMS: Bz-Phe-Val-Arg-AMCRESEARCH AREA: HematologyREFERENCES:L. Svendsen, et al., Folia Haematol, 98, 446 (1972)R. Lottenberg, et al., Meth. Enz., 80, 341 (1981)