Learn More
Cayman Chemical Thrombin (human), 250 U
SDP
Catalog No. 502952449
Encompass_Preferred
Source: Human blood; all source material tested and found non-reactive to HTLV-III and HBsAg using FDA approved assay methods • Mr: 38 kDa
Catalog No. 50-295-2449 Supplier Cayman Chemical Supplier No. 13188250 U
Source: Human blood; all source material tested and found non-reactive to HTLV-III and HBsAg using FDA approved assay methods • Mr: 38 kDa