Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Thrombo-Wellcotest™ Rapid Latex Agglutination Test

Catalog No. R30852301 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3 mL/Vial
Type:
Latex
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
R30852301 3 mL/Vial Latex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R30852301 Supplier Thermo Scientific™ Supplier No. R30852301

missing translation for 'validMainframeMsg'

Rapidly detect fibrin and fibrinogen degradation products in human serum and urine by latex agglutination.

Carry out semi-quantitative detection of fibrin and fibrinogen degradation products in human serum and urine using Thermo Scientific™ Thrombo-Wellcotest™ Rapid Latex Agglutination Test.

Use this simple, rapid, semi-quantitative test for the presence of fibrin/fibrinogen degredation products (FDP) in human serum and urine samples.

The test is designed as a slide agglutination method in which one drop of sample and one drop of latex suspension are mixed. A distinct agglutination pattern at the end of the test period indicates the presence of 2μg/ml FDP or more in the sample under test. By assaying samples at two different dilutions, an approximate FDP value can be determined.

Capable of detecting the presence of all the major breakdown products of fibrin or fibrinogen. The levels measured by this latex test correlate well with results obtained with a haemagglutination-inhibition immunoassay.

The following products are available for use with Thrombo-Wellcotest Test Kit and are sold separately:

  • Thrombo-Wellcotest Latex (Part No. R30852301)
  • Thrombo-Wellcotest Collection Tubes (Part No. R30853001)—Glass tubes contain soya bean trypsin inhibitor (approximately 3600 NF units/tube) and Bothrops atrox venom (>10μg/tube) for the collection of 2mL whole blood or urine.
  • Thrombo-Wellcotest Glycine Saline Buffer (Part No. R30853101)—Glycine saline buffer is provided as a replacement item in instances where multiple tests have exhausted the supply of this component.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Format Vial
Type Latex
Quantity 3 mL/Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.