Learn More
Biomatik Corporation Thrombomodulin (Tm) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU30554
Encompass_Preferred
Thrombomodulin (Tm) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Thrombomodulin (TM). The Thrombomodulin (Tm) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Ala19~Ser515. Applications: WB, IHC, ICC, IP. Target Synonyms: Thrombomodulin (TM). Accession Number: P07204. Gene ID: 7056. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6308