Learn More
Pacific Hemostasis™ Thromboplastin-DS, 10 x 10mL
Detect acquired or congenital disorders affecting the extrinsic and common pathways.
Description
Detect acquired or congenital disorders affecting the extrinsic and common pathways with Thermo Scientific™ Pacific Hemostasis™ Prothrombin Time (PT) Reagents. Widely accepted as a means of monitoring patients on oral anticoagulant therapy such as Warfarin/Coumadin, reagents are manufactured with up to three years extended shelf life and have excellent lot-to-lot consistency.
Features:- Compatible with automated, semi-automated or manual systems
- Reconstitution stability: 7 days at 2° to 8°C (36° to 46°F)
- Dried rabbit brain thromboplastin
Thromboplastin-D with CaCl2 (Product No. 10-0356, 10-0357):
- Target ISI value of 1.7 to 2.2
- Expected normal values of 12 to 15 seconds
Thromboplastin-DS (Product No. 10-0362, 10-0354):
- Target ISI value of 1.0 to 1.3
- Expected normal values of 12 to 15 seconds
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.