Learn More
Siemens Healthineers Thromborel™ S (US)
Catalog No. 23044775 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 10 mL
Catalog No. Quantity
23-044-775 10 x 10 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-775 Supplier Siemens Healthineers Supplier No. 10873565
Add to Cart
Add to Cart
This item is not returnable. View return policy
Determination of prothrombin time (PT) and of the activity of coagulation factors II, V, VII and X
Thromborel™ S Reagent is prepared from human placental tissue factor combined with calcium chloride and stabilizers. The reagent provides a rapid and sensitive screening test for coagulation disorders within the extrinsic coagulation system (factors II, V, VII, and X). Because of its high sensitivity to these coagulation factors, the reagent is suitable for monitoring oral anticoagulant therapy.
Thromborel™ S Reagent exhibits good correlation with the WHO international reference thromboplastin preparation. With Thromborel™ S Reagent and the appropriate deficient plasma, it is possible to determine activity of coagulation factors II, V, VII, and X. The reagent differentiates abnormal plasmas, even in the mildly pathological range.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Prothrombin Time (PT) Testing
Quantity 10 x 10 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov