Learn More
Sigma Aldrich Fine Chemicals Biosciences Thrombospondin 1 human rec
Catalog No. 502904859
Encompass
Recombinant human thrombospondin-1 is expressed in HEK293 cells as a glycoprotein with a calculated moleculaer mass of 127.5 kDa. This protein is manufactured in human cells using an all-human production system with no serum. The human cells expression system allows human-like glycosylation and folding and often supports better stability of the protein in culture.
Catalog No. 50-290-4859 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. ECM00250UG
Recombinant human thrombospondin-1 is expressed in HEK293 cells as a glycoprotein with a calculated moleculaer mass of 127.5 kDa. This protein is manufactured in human cells using an all-human production system with no serum. The human cells expression system allows human-like glycosylation and folding and often supports better stability of the protein in culture.