Learn More
Haematologic Technologies Inc THROMBOSPONDIN 100UG
Supplier:  Haematologic Technologies Inc HCTP0200
Encompass_Preferred
Platelet protein, Human Thrombospondin, 100 micrograms, Purified from platelets, Mode of action: platelet adherence/aggregation, cell-matrix interactions, Research use only, Supplied in 50% (v/v) glycerol/water, Store at -20 Celsius.
Catalog No. 50-883-313