Learn More
Diagnostic Biosystems THROMBOSPONDIN Thrombospondin
Supplier:  Diagnostic Biosystems MOB31501
Encompass_Preferred
This antibody is specific to 450 kDa (non-reduced) and 170 to 180 kD (reduced) form of thrombospondin. Thrombospondin is a protein from platelet α granules. It is secreted at sites of platelet activation and aggregation and is involved in the differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
Catalog No. 50-240-7488