Learn More
ACROBiosystems ACROBiosystems Human Thrombospondin-2 Protein, His Tag
Catalog No. 501957127
Encompass_Preferred
Recombinant Protein;1MG;Human Thrombospondin-2, His Tag (TH2-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 19 - Ile 1172 (Accession # P35442-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 129.7 kDa. The protein migrates as 160 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7127 Supplier ACROBiosystems Supplier No. TH2H52H51MG
Recombinant Protein;1MG;Human Thrombospondin-2, His Tag (TH2-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 19 - Ile 1172 (Accession # P35442-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 129.7 kDa. The protein migrates as 160 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.