Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thromboxane A Synthase Antibody (YA2670) | 100 UL
SDP

Catalog No. 5000445139
Encompass_Preferred

Thromboxane A Synthase Antibody (YA2670) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed for research use. It targets Thromboxane A Synthase, an enzyme crucial for converting prostaglandin H2 into thromboxane A2, a key regulator of blood vessel constriction and platelet aggregation. This antibody is suitable for applications such as Western Blotting.

  • Rabbit-derived recombinant monoclonal antibody
  • Targets thromboxane A synthase
  • Reacts with human and mouse species
  • Immunogen is a synthesized peptide from human thromboxane synthase
  • Isotype is IgG
  • Appearance is liquid
  • Formulated in phosphate buffered saline with BSA, sodium azide, and glycerol
  • Storage at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Applications include Western Blot (WB) at 1:500-1:1000 dilution

Catalog No. 50-004-45139 Supplier Medchemexpress LLC Supplier No. HYP82925100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Thromboxane A Synthase Antibody (YA2670) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed for research use. It targets Thromboxane A Synthase, an enzyme crucial for converting prostaglandin H2 into thromboxane A2, a key regulator of blood vessel constriction and platelet aggregation. This antibody is suitable for applications such as Western Blotting.

  • Rabbit-derived recombinant monoclonal antibody
  • Targets thromboxane A synthase
  • Reacts with human and mouse species
  • Immunogen is a synthesized peptide from human thromboxane synthase
  • Isotype is IgG
  • Appearance is liquid
  • Formulated in phosphate buffered saline with BSA, sodium azide, and glycerol
  • Storage at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Applications include Western Blot (WB) at 1:500-1:1000 dilution

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.