Learn More
Medchemexpress LLC Thromboxane A Synthase Antibody (YA2670) | 50 UL
SDP
Supplier:  Medchemexpress LLC HYP8292550UL
Encompass_Preferred
This rabbit-derived, non-conjugated IgG monoclonal antibody targets Thromboxane A Synthase. Designed for research use only, it catalyzes the conversion of prostaglandin H2 (PGH2) to thromboxane A2 (TXA2), a potent inducer of blood vessel constriction and platelet aggregation. It plays a role in pathophysiological processes including hemostasis, cardiovascular disease, and stroke.
  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets thromboxane A synthase
  • Predicted molecular weight: 61 kDa
  • Observed molecular weight: 54 kDa
  • For research use only
  • Catalyzes conversion of prostaglandin H2 (PGH2) to thromboxane A2 (TXA2)
  • Potent inducer of blood vessel constriction and platelet aggregation
  • Involved in hemostasis, cardiovascular disease, and stroke
Catalog No. 50-004-44193