Learn More
Cayman Chemical Thromboxane B2 CCR2RAR 5mg
SDP
Catalog No. 502941540
Encompass_Preferred
An inhibitor of ACC1 and ACC2 (IC50s 2 1 and 6 1 nM respectively for the human enzymes) inhibits ACC dimerization selective for ACC over 101 enzymes receptors growth factors transporters and ion channels up to a concentration of 10 UM reduces fatty acid synthesis in HepG2 cells (EC50s 66 and 9 nM in 10 FBS and serum-free media respectively) reduces hepatic steatosis improves glucose-stimulated insulin secretion and reduces hemoglobin A1c levels by 0 9 in Zucker diabetic rats
Catalog No. 50-294-1540 Supplier Cayman Chemical Supplier No. 28423 5MG
An inhibitor of ACC1 and ACC2 (IC50s 2 1 and 6 1 nM respectively for the human enzymes) inhibits ACC dimerization selective for ACC over 101 enzymes receptors growth factors transporters and ion channels up to a concentration of 10 UM reduces fatty acid synthesis in HepG2 cells (EC50s 66 and 9 nM in 10 FBS and serum-free media respectively) reduces hepatic steatosis improves glucose-stimulated insulin secretion and reduces hemoglobin A1c levels by 0 9 in Zucker diabetic rats