Learn More
Cayman Chemical Thromboxane B2 NdB2564T 1g
SDP
Catalog No. 502967117
Encompass_Preferred
An internal standard for the quantification of JWH 081 N-pentanoic acid metabolite by GC- or LC-MS
Catalog No. 50-296-7117 Supplier Cayman Chemical Supplier No. 17443 1G
An internal standard for the quantification of JWH 081 N-pentanoic acid metabolite by GC- or LC-MS