Learn More
Biomatik Corporation Thromboxane B2 (TXB2) Instant ELISA Kit
Supplier:  Biomatik Corporation EKU10375
Encompass_Preferred
Thromboxane B2 (TXB2) Instant ELISA Kit for detecting TXB2 in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 18.52-1,500pg/ml. Sensitivity: 7.14pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 1h, 10min. Target Synonyms: TX-B2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8334