Learn More
Cayman Chemical Thromboxane B2, Purity: ≥98%, CAS: 54397-85-2, 10 mg
SDP
Catalog No. 502983437
Encompass_Preferred
An inactive metabolite of TXA2; serum levels positively correlate with platelet COX-1 activation; urinary levels reflect intrarenal TXA2 synthesis
Catalog No. 50-298-3437 Supplier Cayman Chemical Supplier No. 1903010 MG
An inactive metabolite of TXA2; serum levels positively correlate with platelet COX-1 activation; urinary levels reflect intrarenal TXA2 synthesis