Learn More
Cayman Chemical Thromboxn B2-bIotIn 100UG
SDP
Catalog No. 502892577
Encompass_Preferred
An affinity probe which allows TXB2 to be detected through an interaction with the biotin ligand
Catalog No. 50-289-2577 Supplier Cayman Chemical Supplier No. 9002298100 UG
An affinity probe which allows TXB2 to be detected through an interaction with the biotin ligand