Learn More
Cayman Chemical Thromboxn B2-bIotIn 500UG
SDP
Catalog No. 502893641
Encompass_Preferred
An affinity probe which allows TXB2 to be detected through an interaction with the biotin ligand
Catalog No. 50-289-3641 Supplier Cayman Chemical Supplier No. 9002298500 UG
An affinity probe which allows TXB2 to be detected through an interaction with the biotin ligand