Learn More
Cayman Chemical Thromboxn B2-d4 100ug
SDP
Catalog No. 502947326
Encompass_Preferred
An internal standard for the quantification of TXB2 by GC- or LC-MS
Catalog No. 50-294-7326 Supplier Cayman Chemical Supplier No. 319030100 UG
An internal standard for the quantification of TXB2 by GC- or LC-MS