Learn More
Cayman Chemical Thromboxn B2-d4 50ug
SDP
Catalog No. 502949439
Encompass_Preferred
An internal standard for the quantification of TXB2 by GC- or LC-MS
Catalog No. 50-294-9439 Supplier Cayman Chemical Supplier No. 31903050 UG
An internal standard for the quantification of TXB2 by GC- or LC-MS