Learn More
Cayman Chemical Thromboxn B3 100ug
SDP
Catalog No. 502983446
Encompass_Preferred
A uremic toxin and metabolite of tryptophan activates AhR in HepG2 40/6 hepatoma cells (EC50 12 1 nM in a reporter assay) inhibits OAT1 and OAT3 (Kis 34 2 and 74 4 UM respectively for the rat transporters) in S2 proximal tubule cells increases superoxide anion and nitric oxide levels in isolated human mononuclear blood cells at 0 2 and 1 mM increases serum creatinine and BUN levels in the 5/6 nephrectomized rat model of chronic renal failure at 50 mg/kg
Catalog No. 50-298-3446 Supplier Cayman Chemical Supplier No. 19990100 UG
A uremic toxin and metabolite of tryptophan activates AhR in HepG2 40/6 hepatoma cells (EC50 12 1 nM in a reporter assay) inhibits OAT1 and OAT3 (Kis 34 2 and 74 4 UM respectively for the rat transporters) in S2 proximal tubule cells increases superoxide anion and nitric oxide levels in isolated human mononuclear blood cells at 0 2 and 1 mM increases serum creatinine and BUN levels in the 5/6 nephrectomized rat model of chronic renal failure at 50 mg/kg