Learn More
Takara Bio ThruPLEX DNA-Seq Kit, 24 Reactions
SDP
Catalog No. NC1675744
Encompass
The SMARTer ThruPLEX DNA-Seq Kit builds on the innovative ThruPLEX chemistry to generate high-complexity DNA libraries from as little as 50 pg of DNA. Single index, dual index, and unique dual index kits are available and must be purchased separately. This product contains reagents for 24 reactions.
Catalog No. NC1675744 Supplier Takara Bio Supplier No. R400674
The SMARTer ThruPLEX DNA-Seq Kit builds on the innovative ThruPLEX chemistry to generate high-complexity DNA libraries from as little as 50 pg of DNA. Single index, dual index, and unique dual index kits are available and must be purchased separately. This product contains reagents for 24 reactions.