Learn More
Takara Bio THRUPLEX DNA-SEQ KIT 48RXNS
SDP
Catalog No. NC1856611
Encompass
The ThruPLEX DNA-Seq Kit builds on the innovative ThruPLEX chemistry to generate high-complexity DNA libraries from as little as 50 pg of DNA. Single index, dual index, and unique dual index kits are available and must be purchased separately. This product contains reagents for 48 reactions.Our products are to be used for Research Use Only. TAKARA BIO USA INC mfg# R400675
Catalog No. NC1856611 Supplier Takara Bio Supplier No. R400675
The ThruPLEX DNA-Seq Kit builds on the innovative ThruPLEX chemistry to generate high-complexity DNA libraries from as little as 50 pg of DNA. Single index, dual index, and unique dual index kits are available and must be purchased separately. This product contains reagents for 48 reactions.Our products are to be used for Research Use Only. TAKARA BIO USA INC mfg# R400675