Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ THSD1 Antibody Pair [HRP]
SDP

Catalog No. NB010189
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010189 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010189 Supplier Novus Biologicals™ Supplier No. NBP27957515PLATES

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Polyclonal/Monoclonal Antibody Pair

specifically detects in Mouse samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 62.5 pg/mL
Assay Sensitivity 62.5-4000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Mouse THSD1.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias thrombospondin type-1 domain-containing protein 1, thrombospondin, type I, domain 1, thrombospondin, type I, domain containing 1,4833423O18Rik, TMTSPMGC74971, Transmembrane molecule with thrombospondin module, UNQ3010
Gene ID (Entrez) 55901
Gene Symbol THSD1
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.25 mg/mL rabbit anti-THSD1 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.5 mg/mL of rabbit anti-THSD1 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 95 ng of recombinant THSD1.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer
    Storage Requirements Storage is content dependent.
    Target THSD1
    Technique 1. Capture Antibody: Dilute to a working concentration of 1 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 4 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.