Learn More
Aviva Systems Biology Corp THTPA Antibody - N-terminal region (ARP62928_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62928_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against THTPA. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human THTPA. Alternate names: MGC2652, THTP, THTPASEApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-809