Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Thulium(III) chloride hexahydrate

Catalog No. 2046681G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-466-81G 1 g
20-466-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-466-81G Supplier Sigma Aldrich Supplier No. 2046681G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

TmCl3 has been used as a precursor to Tm during the synthesis of NaYF4:Yb,Tm upconversion nanocrystal-semiconductor hybrid structures, which finds use in enhanced NIR photocatalysis.

Specifications

CAS 1331-74-4
Molecular Formula Cl3Tm -+ 6H2O
Linear Formula TmCl3 -+ 6H2O
Quantity 1 g
MDL Number MFCD00149878
Synonym Thulium trichloride
Molecular Weight (g/mol) 383.38
Percent Purity 99.99% trace metals basis
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.