Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thulium(III) oxide, 99.9% (REO)

Catalog No. AA3627904
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3627904 2 g
AA3627909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3627904 Supplier Thermo Scientific Chemicals Supplier No. 03627904
Add to Cart

CAS: 12036-44-1 | O3Tm2 | 385.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-44-1
Molecular Formula O3Tm2
Molecular Weight (g/mol) 385.87
MDL Number MFCD00011285
InChI Key ZIKATJAYWZUJPY-UHFFFAOYSA-N
IUPAC Name dithulium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Tm+3].[Tm+3]

Specifications

Melting Point 2425°C
Color Green
Physical Form Sintered Lump
Quantity 2 g
Linear Formula Tm2O3
Merck Index 14,9394
Formula Weight 385.87
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Thulium(III) oxide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 234-851-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.