Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thulium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific™

Catalog No. AA40565NC
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40565NC 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40565NC Supplier Thermo Scientific Chemicals Supplier No. 40565NC

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14014-10-9
Molecular Formula Cl3O12Tm
Molecular Weight (g/mol) 467.272
MDL Number MFCD00049609
InChI Key JNNWMYZIHRZEKX-UHFFFAOYSA-K
Synonym thulium 3+ perchlorate, thulium iii perchlorate, perchloric acid,thulium 3+ salt 8ci,9ci, acmc-1bs9x, perchloric acid thulium iii salt, thulium 3+ triperchlorate, thulium iii perchlorate w/w aqueous solution, reagent grade
PubChem CID 15335706
IUPAC Name thulium(3+);triperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Tm+3]

Specifications

Physical Form Liquid
Quantity 10 g
UN Number UN3211
Formula Weight 467.28
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Density 1.51 g/mL
Chemical Name or Material Thulium(III) perchlorate
Hazard Category H271-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
EINECSNumber 237-827-3
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.