Learn More
Description
- Clipper-style thumb mechanism enables controlled, one-handed ear notch placement with minimal restraint required for the research animal
- 2 mm punch diameter produces clean, consistent notches that conform to standard small-animal ear-notching identification protocols
- Premium stainless steel construction is autoclave-compatible, allowing full sterilization between uses in accordance with facility biosafety requirements
- Precision-ground cutting edges deliver a clean punch with minimal tissue compression or tearing for accurate, readable animal identification marks
- Compact and lightweight for easy single-hand use during high-throughput colony management or routine ear-notching sessions
- Suited for mice, rats, and other small research animals commonly used in biomedical and preclinical research settings
Specifications
Specifications
| Diameter (Metric) | 2 mm |
| For Use With (Application) | General Lab Procedures |
| Length (English) | 3.25 in. |
| Animal | Small Animal |
| Type | Ear Punch |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.