Learn More
Sklar Instruments Econo Thumb Tissue Forceps, Sklar, 5-1/2 in., 1x2 Teeth, Straight, Smooth Jaws, Stainless Steel. PK/12.
SDP
Catalog No. 501437680 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Thumb Tissue Forceps, Sklar, 5-1/2 in., 1x2 Teeth, Straight, Smooth Jaws, Stainless Steel. Econo Eye Tissue Forceps are most commonly used in ophthalmic procedures for grasping and holding tissue. This instrument is available in several different variations. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with 1x2 teeth and a length of 5-1/4 inches. It is sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7680 Supplier Sklar Instruments Supplier No. 21762
Econo Thumb Tissue Forceps, Sklar, 5-1/2 in., 1x2 Teeth, Straight, Smooth Jaws, Stainless Steel. Econo Eye Tissue Forceps are most commonly used in ophthalmic procedures for grasping and holding tissue. This instrument is available in several different variations. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with 1x2 teeth and a length of 5-1/4 inches. It is sold in boxes of 12.