Learn More
Aviva Systems Biology Corp THUMPD2 Antibody - N-terminal region (ARP48937_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48937_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against THUMPD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human THUMPD2. Alternate names: C2orf8, MGC2454ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59965