Learn More
American Research Products Inc THUNDER™ Extreme Phospho-ERK1/2 TR-FRET Assay Kit, High-Sensitivity Detection, No-Wash Homogeneous Format, 10000 Points
Catalog No. NC3592519
Encompass
The THUNDER™ Extreme Phospho-ERK1/2 (T202/Y204) TR-FRET Cell Signaling Assay Kit provides high-sensitivity detection of phosphorylated ERK1/2 in cell lysates. Its no-wash, homogeneous protocol allows for streamlined high-throughput screening, making it an efficient choice in academic and drug discovery applications. Unique compatibility with both adherent and suspension cells enhances its versatility, establishing it as a cost-effective alternative to traditional assays.
Catalog No. NC3592519 Supplier American Research Products Inc Supplier No. KITXTMERKP10000
Add to Cart
Add to Cart
Request a Quote
The THUNDER™ Extreme Phospho-ERK1/2 (T202/Y204) TR-FRET Cell Signaling Assay Kit provides high-sensitivity detection of phosphorylated ERK1/2 in cell lysates. Its no-wash, homogeneous protocol allows for streamlined high-throughput screening, making it an efficient choice in academic and drug discovery applications. Unique compatibility with both adherent and suspension cells enhances its versatility, establishing it as a cost-effective alternative to traditional assays.