Learn More
American Research Products Inc Thunder Extreme Phospho-ERK1/2 Assay Kit, High-Sensitivity TR-FRET Immunoassay, Compatible With Adherent and Suspension Cells
Catalog No. NC3441072
Encompass
The THUNDER™ Extreme Phospho-ERK1/2 (T202/Y204) TR-FRET Cell Signaling Assay Kit offers high-sensitivity measurement of phosphorylated ERK1/2 in cell lysates. Its streamlined "add-incubate-measure" workflow eliminates wash steps, providing robust results for high-throughput screening applications. Compatible with various cell types, this kit is distinguished by its superior signal-to-background ratios through proprietary TR-FRET technology, ensuring reliable performance.
Catalog No. NC3441072 Supplier American Research Products Inc Supplier No. KITXTMERKP5000
Add to Cart
Add to Cart
Request a Quote
The THUNDER™ Extreme Phospho-ERK1/2 (T202/Y204) TR-FRET Cell Signaling Assay Kit offers high-sensitivity measurement of phosphorylated ERK1/2 in cell lysates. Its streamlined "add-incubate-measure" workflow eliminates wash steps, providing robust results for high-throughput screening applications. Compatible with various cell types, this kit is distinguished by its superior signal-to-background ratios through proprietary TR-FRET technology, ensuring reliable performance.