Learn More
Gold Standard Diagnostics ThunderBolt™ ELISA Analyzer
SDP
Catalog No. 23700160
Change view
Click to view available options
Type:
Automated Enzyme Immunoassay Analyzer
Catalog No. Type
23-700-160 Automated Enzyme Immunoassay Analyzer
1 options
This item is not returnable. View return policy
Catalog No. 23-700-160 Supplier Gold Standard Diagnostics Supplier No. 00300
Add to Cart
Add to Cart
This item is not returnable. View return policy
A compact, fully automated, open platform designed to simplify ELISA and CLIA testing with efficiency.
  • Runs ELISA and Chemiluminescence (CLIA) assays
  • Walk-away instrument to simplify laboratory experience
  • Helps increase productivity in the lab
  • Designed to reduce reagent waste
Specifications
Type Automated Enzyme Immunoassay Analyzer
Capacity 12 racks of 16 samples, up to 192 samples
Certifications/Compliance CE
Wavelength 5, 450, 490, 550 and 630 nm
Electrical Requirements 100/265 V, 50/60 HZ, 120 W
Dimensions (L x W x H) 57 x 64 x 45 cm