Learn More
Cayman Chemical THUNDERHumn GMCSF TRFRET 96Wel
SDP
Catalog No. 502894000
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human GM-CSF in cell culture supernatants
Catalog No. 50-289-4000 Supplier Cayman Chemical Supplier No. 50030796 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human GM-CSF in cell culture supernatants