Learn More
Cayman Chemical THUNDERHumn IL-1 TR-FRET 96Wel
SDP
Catalog No. 502893190
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-1β (IL-1F2) in cell culture supernatants
Catalog No. 50-289-3190 Supplier Cayman Chemical Supplier No. 50023096 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-1β (IL-1F2) in cell culture supernatants