Learn More
Cayman Chemical THUNDERHumn IL1 TRFRET 4800Wel
SDP
Catalog No. 502895999
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-1β (IL-1F2) in cell culture supernatants
Catalog No. 50-289-5999 Supplier Cayman Chemical Supplier No. 5002304800 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-1β (IL-1F2) in cell culture supernatants