Learn More
Cayman Chemical THUNDERHumn IL1 TRFRET 9600Wel
SDP
Catalog No. 502895509
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-1β (IL-1F2) in cell culture supernatants
Catalog No. 50-289-5509 Supplier Cayman Chemical Supplier No. 5002309600 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-1β (IL-1F2) in cell culture supernatants