Learn More
Cayman Chemical THUNDERHumn IL10 TRFRET 480Wel
SDP
Catalog No. 502895550
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-10 (CSIF) in cell culture supernatants
Catalog No. 50-289-5550 Supplier Cayman Chemical Supplier No. 500302480 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-10 (CSIF) in cell culture supernatants