Learn More
Cayman Chemical THUNDERHumn IL10 TRFRET 96
SDP
Catalog No. 502896531
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-10 (CSIF) in cell culture supernatants
Catalog No. 50-289-6531 Supplier Cayman Chemical Supplier No. 50030296 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-10 (CSIF) in cell culture supernatants