Learn More
Cayman Chemical THUNDERHumn IL11 TRFRET 480Wel
SDP
Catalog No. 502892373
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL‑11 in cell culture supernatants
Catalog No. 50-289-2373 Supplier Cayman Chemical Supplier No. 500235480 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL‑11 in cell culture supernatants