Learn More
Cayman Chemical THUNDERHumn IL12IL23 p4 480Wel
SDP
Catalog No. 502892545
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-12/IL-23 p40 (IL-12B; NKFS; CLMF) in cell culture supernatants
Catalog No. 50-289-2545 Supplier Cayman Chemical Supplier No. 500236480 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-12/IL-23 p40 (IL-12B; NKFS; CLMF) in cell culture supernatants