Learn More
Cayman Chemical THUNDERHumn IL17A TRFRET 96Wel
SDP
Catalog No. 502892672
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-17A (IL-17; CTLA-8) in cell culture supernatants
Catalog No. 50-289-2672 Supplier Cayman Chemical Supplier No. 50029296 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-17A (IL-17; CTLA-8) in cell culture supernatants