Learn More
Cayman Chemical THUNDERHumn IL18 TRFRET 480Wel
SDP
Catalog No. 502895202
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL‑18 (IL-1F4) in cell culture supernatants
Catalog No. 50-289-5202 Supplier Cayman Chemical Supplier No. 500304480 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL‑18 (IL-1F4) in cell culture supernatants