Learn More
Cayman Chemical THUNDERHumn IL2 TRFRET B 96Wel
SDP
Catalog No. 502895716
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-2 in cell culture supernatants
Catalog No. 50-289-5716 Supplier Cayman Chemical Supplier No. 50023196 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-2 in cell culture supernatants